Assay Design
Gene Transcripts
- Gene Symbol
- Slc7a8
- Entrez Gene ID
-
84551
- Gene Name
- solute carrier family 7 (amino acid transporter light chain, L system), member 8
- Gene Aliases
-
Lat2, Lat4
- Chromosome Location
-
Chr.15: 33369245 - 33428942 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053442.1 |
NP_445894.1 |
10 - 11 |
2041 |
75 |
GenBank mRNA |
AB024400.1 |
- |
10 - 11 |
2041 |
75 |
View Full Product Details