Assay Design



Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
290235
Gene Name
NOP9 nucleolar protein
Gene Aliases
RGD1308396
Chromosome Location
Chr.15: 34431357 - 34439861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106040.1 NP_001099510.1 1 - 2 317 86

View Full Product Details