Assay Design



Gene Transcripts

Gene Symbol
Zfhx2
Entrez Gene ID
305888
Gene Name
zinc finger homeobox 2
Gene Aliases
ZFH
Chromosome Location
Chr.15: 33719313 - 33752665 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098803.1 NP_001092273.1 2 - 3 2386 64
XM_008770677.1 XP_008768899.1 1 - 2 4161 64
XM_008770678.1 XP_008768900.1 1 - 2 4161 64

View Full Product Details