Assay Design



Gene Transcripts

Gene Symbol
Psme1
Entrez Gene ID
29630
Gene Name
proteasome (prosome, macropain) activator subunit 1
Gene Aliases
-
Chromosome Location
Chr.15: 34256071 - 34258947 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017264.3 NP_058960.2 6 - 7 497 71
GenBank mRNA BC060574.1 - 6 - 7 501 71
D45249.1 - 6 - 7 421 71
FQ216423.1 - 6 - 7 498 71
FQ219304.1 - 6 - 7 500 71
FQ220175.1 - 6 - 7 446 71
FQ224986.1 - 6 - 7 444 71
FQ228562.1 - - 452 71
FQ230622.1 - 6 - 7 444 71
FQ233032.1 - 6 - 7 450 71
FQ233635.1 - 6 - 7 450 71
FQ233859.1 - 6 - 7 446 71

View Full Product Details