Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017264.3 | NP_058960.2 | 7 - 8 | 562 | 141 |
GenBank mRNA | BC060574.1 | - | 7 - 8 | 566 | 141 |
FQ216423.1 | - | 7 - 8 | 563 | 141 | |
FQ220175.1 | - | 7 - 8 | 511 | 141 | |
FQ224986.1 | - | 7 - 8 | 509 | 141 | |
FQ226855.1 | - | 7 - 8 | 515 | 141 | |
FQ228562.1 | - | - | 517 | 141 | |
FQ230622.1 | - | 7 - 8 | 509 | 141 | |
FQ233032.1 | - | 7 - 8 | 515 | 141 | |
FQ233635.1 | - | 7 - 8 | 515 | 141 | |
FQ233859.1 | - | 7 - 8 | 511 | 141 |