Assay Design



Gene Transcripts

Gene Symbol
Tbca
Entrez Gene ID
366995
Gene Name
tubulin folding cofactor A
Gene Aliases
-
Chromosome Location
Chr.2: 24449497 - 24505353 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013245.1 NP_001013263.1 2 - 3 186 67
GenBank mRNA BC058155.1 - 2 - 3 186 67

View Full Product Details