Assay Design



Gene Transcripts

Gene Symbol
Poc5
Entrez Gene ID
294667
Gene Name
POC5 centriolar protein
Gene Aliases
RGD1310597
Chromosome Location
Chr.2: 27000303 - 27029605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025647.1 NP_001020818.1 6 - 7 842 105
XM_006231821.1 XP_006231883.1 4 - 5 538 105
XM_008760664.1 XP_008758886.1 3 - 4 583 105
XM_008760665.1 XP_008758887.1 4 - 5 461 105
GenBank mRNA BC097489.1 - 6 - 7 842 105

View Full Product Details