Assay Design



Gene Transcripts

Gene Symbol
Myog
Entrez Gene ID
29148
Gene Name
myogenin
Gene Aliases
-
Chromosome Location
Chr.13: 51126459 - 51129048 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017115.2 NP_058811.2 1 - 2 505 94
GenBank mRNA AF054894.1 - 1 - 2 471 94
M24393.1 - 1 - 2 504 93

View Full Product Details