Assay Design



Gene Transcripts

Gene Symbol
Fcamr
Entrez Gene ID
681070
Gene Name
Fc receptor, IgA, IgM, high affinity
Gene Aliases
-
Chromosome Location
Chr.13: 47538783 - 47552799 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001060181.4 XP_001060181.2 4 - 5 1014 104
XM_008769511.1 XP_008767733.1 3 - 4 884 104

View Full Product Details