Assay Design



Gene Transcripts

Gene Symbol
Srfbp1
Entrez Gene ID
291469
Gene Name
serum response factor binding protein 1
Gene Aliases
p49/STRAP
Chromosome Location
Chr.18: 47456406 - 47478532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005536.1 NP_001005536.1 3 - 4 287 126
GenBank mRNA BC082077.1 - 3 - 4 287 126

View Full Product Details