Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107379.1 | NP_001100849.1 | 11 - 12 | 3177 | 100 |
XM_006254719.2 | XP_006254781.1 | 11 - 12 | 3163 | 100 | |
XM_006254720.2 | XP_006254782.1 | 11 - 12 | 2914 | 100 | |
XM_006254721.2 | XP_006254783.1 | 11 - 12 | 3008 | 100 |