Assay Design



Gene Transcripts

Gene Symbol
Cd74
Entrez Gene ID
25599
Gene Name
Cd74 molecule, major histocompatibility complex, class II invariant chain
Gene Aliases
INVG34
Chromosome Location
Chr.18: 56071420 - 56080851 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254761.2 XP_006254823.1 7 - 8 918 84
GenBank mRNA X14254.1 - 7 - 8 824 84

View Full Product Details