Assay Design



Gene Transcripts

Gene Symbol
Mbd2
Entrez Gene ID
680172
Gene Name
methyl-CpG binding domain protein 2
Gene Aliases
-
Chromosome Location
Chr.18: 65814026 - 65885115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115025.1 NP_001108497.1 6 - 7 1256 128
GenBank mRNA BC158877.1 - 6 - 7 1256 128

View Full Product Details