Assay Design



Gene Transcripts

Gene Symbol
Csnk1a1
Entrez Gene ID
113927
Gene Name
casein kinase 1, alpha 1
Gene Aliases
-
Chromosome Location
Chr.18: 56887722 - 56910610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053615.1 NP_446067.1 7 - 8 751 130
GenBank mRNA BC127486.1 - - 977 130
U77582.1 - 7 - 8 751 130

View Full Product Details