Assay Design



Gene Transcripts

Gene Symbol
Clec4e
Entrez Gene ID
450223
Gene Name
C-type lectin domain family 4, member E
Gene Aliases
Clecsf9, Mincle
Chromosome Location
Chr.4: 156271087 - 156276243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005897.1 NP_001005897.1 3 - 4 274 146
GenBank mRNA AY363175.1 - 3 - 4 274 146
AY494064.1 - 3 - 4 240 146
AY581049.1 - 3 - 4 240 146

View Full Product Details