Assay Design



Gene Transcripts

Gene Symbol
Ccnd2
Entrez Gene ID
64033
Gene Name
cyclin D2
Gene Aliases
-
Chromosome Location
Chr.4: 159674885 - 159697207 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA D16308.1 - 2 - 3 590 81
FQ227079.1 - 2 - 3 622 81
FQ233117.1 - 2 - 3 622 81
FQ234359.1 - 2 - 3 622 81

View Full Product Details