Assay Design



Gene Transcripts

Gene Symbol
Sspn
Entrez Gene ID
500364
Gene Name
sarcospan
Gene Aliases
RGD1559723
Chromosome Location
Chr.4: 180291389 - 180325652 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109255.1 NP_001102725.1 2 - 3 308 63

View Full Product Details