Assay Design



Gene Transcripts

Gene Symbol
Ltbr
Entrez Gene ID
297604
Gene Name
lymphotoxin beta receptor (TNFR superfamily, member 3)
Gene Aliases
-
Chromosome Location
Chr.4: 157822838 - 157829291 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008315.1 NP_001008316.1 5 - 6 767 88
XM_006237351.1 XP_006237413.1 5 - 6 817 88
GenBank mRNA BC085880.1 - 5 - 6 767 88

View Full Product Details