Assay Design



Gene Transcripts

Gene Symbol
Nrip2
Entrez Gene ID
689619
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 161719431 - 161729673 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001071392.5 XP_001071392.2 4 - 5 1613 59
XM_006237431.2 XP_006237493.1 4 - 5 1615 59

View Full Product Details