Assay Design
Gene Transcripts
- Gene Symbol
- Foxm1
- Entrez Gene ID
-
58921
- Gene Name
- forkhead box M1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 161685283 - 161697633 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031633.3 |
NP_113821.2 |
5 - 6 |
967 |
67 |
GenBank mRNA |
U83112.1 |
- |
5 - 6 |
967 |
67 |
View Full Product Details