Assay Design



Gene Transcripts

Gene Symbol
Il17ra
Entrez Gene ID
312679
Gene Name
interleukin 17 receptor A
Gene Aliases
Il17r
Chromosome Location
Chr.4: 152995865 - 153018394 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107883.2 NP_001101353.2 4 - 5 525 169

View Full Product Details