Assay Design



Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
64088
Gene Name
sorting nexin 16
Gene Aliases
-
Chromosome Location
Chr.2: 93518796 - 93541076 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022289.3 NP_071625.2 6 - 7 1106 98
GenBank mRNA AF305780.1 - 6 - 7 1092 98
BC061554.1 - 6 - 7 1110 98

View Full Product Details