Assay Design



Gene Transcripts

Gene Symbol
Trim55
Entrez Gene ID
365751
Gene Name
tripartite motif-containing 55
Gene Aliases
Rnf29
Chromosome Location
Chr.2: 104416934 - 104459117 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012218.1 NP_001012218.1 5 - 6 967 110
XM_006232181.2 XP_006232243.1 5 - 6 1005 110
XM_006232182.2 XP_006232244.1 5 - 6 1005 110
GenBank mRNA BC087100.1 - 5 - 6 967 110

View Full Product Details