Assay Design
Gene Transcripts
- Gene Symbol
- Ckmt2
- Entrez Gene ID
-
688698
- Gene Name
- creatine kinase, mitochondrial 2, sarcomeric
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 21422680 - 21452797 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ216242.1 |
- |
4 - 5 |
755 |
66 |
View Full Product Details