Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022538.2 | NP_071983.2 | 5 - 6 | 823 | 163 |
XM_006231949.2 | XP_006232011.1 | 5 - 6 | 1191 | 163 | |
GenBank mRNA | BC061815.1 | - | 5 - 6 | 823 | 163 |
FQ229687.1 | - | 3 - 4 | 418 | 163 | |
JQ013728.1 | - | 5 - 6 | 761 | 163 |