Assay Design



Gene Transcripts

Gene Symbol
Fst
Entrez Gene ID
24373
Gene Name
follistatin
Gene Aliases
FOL1, Fst-288, RATFOL1
Chromosome Location
Chr.2: 46537589 - 46544812 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012561.1 NP_036693.1 5 - 6 954 109
XM_006231953.2 XP_006232015.1 5 - 6 1311 109

View Full Product Details