Assay Design



Gene Transcripts

Gene Symbol
Setd7
Entrez Gene ID
689954
Gene Name
SET domain containing (lysine methyltransferase) 7
Gene Aliases
-
Chromosome Location
Chr.2: 140576188 - 140618405 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109558.1 NP_001103028.1 7 - 8 1012 72

View Full Product Details