Assay Design
Gene Transcripts
- Gene Symbol
- Arhgef26
- Entrez Gene ID
-
310460
- Gene Name
- Rho guanine nucleotide exchange factor (GEF) 26
- Gene Aliases
-
RGD1562559, Sgef
- Chromosome Location
-
Chr.2: 27107276 - 27236336 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_227201.8 |
XP_227201.4 |
4 - 5 |
1871 |
62 |
GenBank mRNA |
FQ218213.1 |
- |
3 - 4 |
516 |
62 |
View Full Product Details