Assay Design



Gene Transcripts

Gene Symbol
Fxr2
Entrez Gene ID
287433
Gene Name
fragile X mental retardation, autosomal homolog 2
Gene Aliases
Fxr2h
Chromosome Location
Chr.10: 56237814 - 56257877 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100647.1 NP_001094117.1 14 - 15 1756 67
GenBank mRNA BC160929.1 - 14 - 15 1756 67

View Full Product Details