Assay Design



Gene Transcripts

Gene Symbol
Krt19
Entrez Gene ID
360626
Gene Name
keratin 19, type I
Gene Aliases
Ka19, Krt1-19
Chromosome Location
Chr.10: 88055843 - 88060560 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY464140.1 - 5 - 6 1021 84
BC088424.1 - 5 - 6 1017 84
BC126075.1 - 5 - 6 1012 84

View Full Product Details