Assay Design



Gene Transcripts

Gene Symbol
Tp53i13
Entrez Gene ID
287550
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
RGD1559818, Trp53i13
Chromosome Location
Chr.10: 62650641 - 62655327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105818.1 NP_001099288.1 6 - 7 1115 134
XM_006246902.2 XP_006246964.1 5 - 6 1142 134
GenBank mRNA BC158678.1 - 6 - 7 1077 134

View Full Product Details