Assay Design
Gene Transcripts
- Gene Symbol
- Stat5a
- Entrez Gene ID
-
24918
- Gene Name
- signal transducer and activator of transcription 5A
- Gene Aliases
-
Stat5
- Chromosome Location
-
Chr.10: 88764732 - 88789060 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017064.1 |
NP_058760.1 |
16 - 17 |
2111 |
162 |
GenBank mRNA |
U24175.1 |
- |
16 - 17 |
2111 |
162 |
View Full Product Details