Assay Design



Gene Transcripts

Gene Symbol
Pkp1
Entrez Gene ID
304822
Gene Name
plakophilin 1
Gene Aliases
-
Chromosome Location
Chr.13: 52705174 - 52753099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107181.1 NP_001100651.1 5 - 6 1245 62
XM_006249884.2 XP_006249946.1 5 - 6 1347 62

View Full Product Details