Assay Design
Gene Transcripts
- Gene Symbol
- Clic6
- Entrez Gene ID
-
304081
- Gene Name
- chloride intracellular channel 6
- Gene Aliases
-
Clic6b
- Chromosome Location
-
Chr.11: 32655653 - 32698004 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176078.2 |
NP_788267.1 |
4 - 5 |
1805 |
133 |
GenBank mRNA |
AY191249.1 |
- |
4 - 5 |
1805 |
133 |
View Full Product Details