Assay Design



Gene Transcripts

Gene Symbol
Thumpd2
Entrez Gene ID
313851
Gene Name
THUMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 3743080 - 3794505 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012108.1 NP_001012108.2 2 - 3 276 150
NM_001163506.1 NP_001156978.1 2 - 3 276 150
XM_006239672.2 XP_006239734.1 2 - 3 280 150
XM_006239673.2 XP_006239735.1 2 - 3 280 150
XM_006239675.2 XP_006239737.1 2 - 3 280 150
XM_006239677.2 XP_006239739.1 2 - 3 280 150
XM_008764440.1 XP_008762662.1 2 - 3 280 150
GenBank mRNA BC089870.1 - 2 - 3 276 150

View Full Product Details