Assay Design



Gene Transcripts

Gene Symbol
Thumpd2
Entrez Gene ID
313851
Gene Name
THUMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 3743080 - 3794505 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163506.1 NP_001156978.1 9 - 10 1198 133

View Full Product Details