Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108333.1 | NP_001101803.1 | 7 - 8 | 1311 | 111 |
XM_006249238.2 | XP_006249300.1 | 8 - 9 | 1406 | 111 | |
XM_006249239.2 | XP_006249301.1 | 8 - 9 | 1158 | 111 | |
XM_006249240.2 | XP_006249302.1 | 8 - 9 | 1395 | 111 | |
XM_008769150.1 | XP_008767372.1 | 8 - 9 | 1402 | 111 | |
GenBank mRNA | BC168697.1 | - | 5 - 6 | 710 | 111 |