Assay Design



Gene Transcripts

Gene Symbol
Tctn1
Entrez Gene ID
304486
Gene Name
tectonic family member 1
Gene Aliases
RGD1566266
Chromosome Location
Chr.12: 39790965 - 39824064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177613.1 NP_001171084.1 11 - 12 1303 62
GenBank mRNA FQ222752.1 - 10 - 11 1383 62

View Full Product Details