Assay Design



Gene Transcripts

Gene Symbol
Tctn1
Entrez Gene ID
304486
Gene Name
tectonic family member 1
Gene Aliases
RGD1566266
Chromosome Location
Chr.12: 39790965 - 39824064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177613.1 NP_001171084.1 13 - 14 1611 57
GenBank mRNA FQ222752.1 - 12 - 13 1691 57

View Full Product Details