Assay Design



Gene Transcripts

Gene Symbol
Iqub
Entrez Gene ID
296936
Gene Name
IQ motif and ubiquitin domain containing
Gene Aliases
RGD1562359, Trs4
Chromosome Location
Chr.4: 51482169 - 51586881 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034130.1 NP_001029302.1 12 - 13 2239 91
GenBank mRNA BC099086.1 - 12 - 13 2239 91
DQ132434.1 - 12 - 13 91

View Full Product Details