Assay Design



Gene Transcripts

Gene Symbol
Fezf1
Entrez Gene ID
500048
Gene Name
Fez family zinc finger 1
Gene Aliases
RGD1560480
Chromosome Location
Chr.4: 50309281 - 50312608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109224.1 NP_001102694.1 2 - 3 1216 62
XM_008762805.1 XP_008761027.1 3 - 4 778 62

View Full Product Details