Assay Design



Gene Transcripts

Gene Symbol
Chchd3
Entrez Gene ID
296966
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 60080538 - 60358562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106588.1 NP_001100058.1 3 - 4 410 77
GenBank mRNA FQ216356.1 - 3 - 4 393 77

View Full Product Details