Assay Design
Gene Transcripts
- Gene Symbol
- Tas2r118
- Entrez Gene ID
-
78980
- Gene Name
- taste receptor, type 2, member 118
- Gene Aliases
-
T2R3, Tas2r16
- Chromosome Location
-
Chr.4: 51002218 - 51003117 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_023994.1 |
NP_076484.1 |
1 - 1 |
842 |
99 |
GenBank mRNA |
AF227141.1 |
- |
1 - 1 |
842 |
99 |
View Full Product Details