Assay Design



Gene Transcripts

Gene Symbol
RGD1312005
Entrez Gene ID
291580
Gene Name
similar to DD1
Gene Aliases
-
Chromosome Location
Chr.18: 54108471 - 54123061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006254754.2 XP_006254816.1 1 - 2 341 111

View Full Product Details