Assay Design
Gene Transcripts
- Gene Symbol
- Ccdc88b
- Entrez Gene ID
-
293698
- Gene Name
- coiled-coil domain containing 88B
- Gene Aliases
-
RGD1311475
- Chromosome Location
-
Chr.1: 222102436 - 222118479 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_215206.9 |
XP_215206.6 |
20 - 21 |
3567 |
99 |
View Full Product Details