Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003749042.3 | XP_003749090.1 | 2 - 3 | 466 | 68 |
XM_003749043.3 | XP_003749091.1 | 2 - 3 | 429 | 68 | |
XM_006230934.1 | XP_006230996.1 | 2 - 3 | 542 | 68 | |
XM_006230935.2 | XP_006230997.1 | 2 - 3 | 412 | 68 | |
XM_006230936.1 | XP_006230998.1 | 2 - 3 | 413 | 68 | |
XM_006230937.2 | XP_006230999.1 | 2 - 3 | 361 | 68 | |
XM_008760196.1 | XP_008758418.1 | 1 - 2 | 850 | 68 | |
GenBank mRNA | BC166555.1 | - | 2 - 3 | 434 | 68 |