Assay Design
Gene Transcripts
- Gene Symbol
- Ctbp2
- Entrez Gene ID
-
81717
- Gene Name
- C-terminal binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 205001354 - 205030565 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053335.1 |
NP_445787.1 |
7 - 8 |
|
104 |
GenBank mRNA |
AF222712.1 |
- |
7 - 8 |
|
104 |
View Full Product Details