Assay Design



Gene Transcripts

Gene Symbol
Catsper1
Entrez Gene ID
689349
Gene Name
cation channel, sperm associated 1
Gene Aliases
-
Chromosome Location
Chr.1: 220715001 - 220723983 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760198.1 XP_008758420.1 6 - 7 1724 123

View Full Product Details