Assay Design



Gene Transcripts

Gene Symbol
Med30
Entrez Gene ID
299905
Gene Name
mediator complex subunit 30
Gene Aliases
Thrap6
Chromosome Location
Chr.7: 92234589 - 92256101 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130539.1 NP_001124011.1 2 - 3 423 168

View Full Product Details