Assay Design



Gene Transcripts

Gene Symbol
Sept3
Entrez Gene ID
56003
Gene Name
septin 3
Gene Aliases
Sep3
Chromosome Location
Chr.7: 123506535 - 123526542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019375.1 NP_062248.1 10 - 11 1049 72
GenBank mRNA AF111179.1 - 10 - 11 1049 72

View Full Product Details